missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ Melting Furnace R7 Controller

These compact melting furnaces for the melting of non-ferrous metals and alloys are one of a kind and have a number of technical advantages. Designed as tabletop models, they can be used for many laboratory applications

Brand:  Nabertherm™ KD0230IRNA

Code : NEW

Additional Details : Weight : 145.00000kg

 View more versions of this product

Product Code. 15541012

  • £5735.00 / Each

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Controller: R7
  • tabletop model
  • Electrical connection: 1-phase
Specifications

Specifications

R7
1.5 L
1300°C
520 x 680 x 660 mm
Crucible A 10
Videos
SDS
Documents
Certificates