Learn More
Thermo Scientific™ Melon Gel™ Monoclonal IgG Purification Kit
Purify mouse monoclonal antibodies from serum, ascites or hybridoma cell culture supernatant with this high-yield, mild monoclonal IgG purification kit.
£38.25 - £1172.00
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10015933 View Documents Thermo Scientific™
45214
Melon Gel Monoclonal IgG Purification Kit £1172.00
1 set View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 07-11-2019
11899360 View Documents Thermo Scientific™
45219
Pierce Ascites Conditioning Reagent £38.25
5mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 31-10-2019
Description
Description
The Melon Gel Monoclonal IgG Purification Kit consists of a large package size of Melon Gel Resin and an optimized protocol for easy and mild purification of monoclonal IgG antibodies from hybridoma culture supernatant or ascites fluid. Unlike typical bind-and-elute affinity methods, Melon Gel Purification is based on the binding of non-IgG proteins from a prepared supernatant or ascites sample, thereby allowing the pure IgG to be recovered in the flow-through (non-bound) fraction. Antibodies are recovered ready for use in assays or subsequent conjugation and other modification steps. There is no need to neutralize or desalt to mitigate the effects of harsh elution buffers. Melon Gel Purification saves time compared to classical Protein A or Protein G methods without compromising the quality of the antibody.
Highlights:
  • Fast – purify antibodies in half the time of required for Protein A or G
  • Efficient – recovery and purity is often better than Protein A or G purification
  • Robust – effective for subclasses of IgG that bind poorly to Protein A or G
  • Gentle – antibodies retain activity because they are not exposed to denaturing conditions
  • Reusable – support can be regenerated and used for up to 6 purifications
  • Compatible – amine-free buffer does not require removal or neutralization
  • Flexible – accommodates a broad range of sample volumes and processing vessels
Includes:
Melon Gel Resin, purification buffer and resin regenerant