missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Megaplex™ PreAmp Primers, Rodent Pool Set v3.0
Megaplex™ PreAmp Primers, Rodent Pool Set v3.0
Brand: Applied Biosystems™ 4444747
Code : 50
Additional Details : Weight : 0.00450kg
Description
For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. With content matched to the Megaplex™ RT Primers, Megaplex™ PreAmp Primers, Rodent Pools Set v3.0 is used with Megaplex™ RT Primers, Rodent Pools Set v3.0. Following reverse transcription using Megaplex™ RT Primers, Megaplex™ PreAmp Primers along with the TaqMan™ PreAmp Master Mix uniformly amplify all microRNAs prior to quantitation by real-time PCR using TaqMan™ MicroRNA Arrays.Specifications
Store at -20°C. | |
qPCR | |
Megaplex™, TaqMan™ | |
Rat, Mouse | |
50 Reactions |
Low | |
PreAmp Primers | |
Preamp Primer | |
50 Reactions |
For Research Use Only. Not for use in diagnostic procedures.