missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550718
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total MED12 / TRAP230 protein.
MED12 Polyclonal antibody specifically detects MED12 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin).Specifications
MED12 | |
Polyclonal | |
Unconjugated | |
Med12 | |
230kDa; activator-recruited cofactor 240 kDa component; ARC240; CAG repeat protein 45; CAGH45; FGS1; HOPA; human opposite paired; Kiaa0192; Med12; MED12S; Mediator complex subunit 12; mediator of RNA polymerase II transcription subunit 12; mediator of RNA polymerase II transcription, subunit 12 homolog; Mopa; OHDOX; OKS; OPA1; OPA-1; OPA-containing protein; OPA-containing protein 1; putative mediator subunit 12; thyroid hormone receptor-associated protein complex 230 kDa component; thyroid hormone receptor-associated protein, 230 kDa subunit; TNRC11; TRAP230; trinucleotide repeat containing 11 (THR-associated protein); trinucleotide repeat containing 11 (THR-associated protein, 230 kDa subunit); trinucleotide repeat-containing gene 11 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59024, 9968 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
A2AGH6, Q93074 | |
Med12 | |
Synthetic peptide corresponding to a region derived from internal residues of human mediator complex subunit 12 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |