missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Measuring Cylinders

Brand:  Duran™ 213951402

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15251719

  • £23.54 / Pack of 2


Estimated Shipment
11-06-2018
Add to basket

Specifications

Specifications

115mm
25mL
±0.5mL
1mL
Videos