missing translation for 'onlineSavingsMsg'
Learn More

MDL 73005EF hydrochloride, Tocris Bioscience™

Potent and selective 5-HT1A partial agonist

Brand:  Tocris (Bio-Techne) 041150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13437623

  • £349.81 / 50mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective 5-HT1A receptor antagonist. Has partial agonist properties.
Specifications

Chemical Identifiers

102908-60-1
394.896
mdl 73005ef hydrochloride, 8-2-2,3-dihydrobenzo b 1,4 dioxin-2-yl methyl amino ethyl-8-azaspiro 4.5 decane-7,9-dione hydrochloride, binospirone hydrochloride, 8-2-1,4-benzodioxan-2-ylmethylamino ethyl-8-azaspiro 4.5 decane-7,9-dione hydrochloride
8-[2-(2,3-dihydro-1,4-benzodioxin-3-ylmethylamino)ethyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
C20H27ClN2O4
UEDYXFOPCMTXEH-UHFFFAOYSA-N
56972149
C1CCC2(C1)CC(=O)N(C(=O)C2)CCNCC3COC4=CC=CC=C4O3.Cl

Specifications

MDL 73005EF hydrochloride
102908-60-1
mdl 73005ef hydrochloride, 8-2-2,3-dihydrobenzo b 1,4 dioxin-2-yl methyl amino ethyl-8-azaspiro 4.5 decane-7,9-dione hydrochloride, binospirone hydrochloride, 8-2-1,4-benzodioxan-2-ylmethylamino ethyl-8-azaspiro 4.5 decane-7,9-dione hydrochloride
C1CCC2(C1)CC(=O)N(C(=O)C2)CCNCC3COC4=CC=CC=C4O3.Cl
394.896
394.9
50mg
C20H27ClN2O4
UEDYXFOPCMTXEH-UHFFFAOYSA-N
8-[2-(2,3-dihydro-1,4-benzodioxin-3-ylmethylamino)ethyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
56972149
>98%
Videos