missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MDL 72832 hydrochloride
Potent 5-HT1A ligand
POA
Specifications
Assay Percent Range | >99% |
---|---|
Chemical Name or Material | 8-[4-(1,4-Benzodioxan-2-ylmethylamino)butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride |
CAS | 113777-40-5 |
Description
A potent ligand at 5-HT1A receptors, with mixed agonist and antagonist properties. An agonist at post-synaptic 5-HT1A receptors.Specifications
>99% | |
8-[4-(1,4-Benzodioxan-2-ylmethylamino)butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride | |
113777-40-5 |
For Research Use Only