Learn More
Tocris Bioscience™ MDL 72832 hydrochloride
Brand:  Tocris Bioscience™ 041210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13447623
  • £92.61 / 10mg
Estimated Shipment date
from Supplier 02-04-2020
Description
Description
A potent ligand at 5-HT1A receptors, with mixed agonist and antagonist properties. An agonist at post-synaptic 5-HT1A receptors.
Specifications
Specifications
8-[4-(1,4-Benzodioxan-2-ylmethylamino)butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride
>99%
Observed MW: 422.95
Soluble to 5 mM in water
113777-40-5
C22H30N2O4.HCl