missing translation for 'onlineSavingsMsg'
Learn More

Mdivi 1, Tocris Bioscience™

Selective dynamin inhibitor; attenuates mitochondrial division and apoptosis

Brand:  Tocris (Bio-Techne) 398250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443924

  • £347.17 / 50mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of Dnm1 GTPase (IC50 = 1 - 10 μM); attenuates Dnm1- and Drp1-mediated mitochondrial division. Inhibits mitochondrial outer membrane permeabilization (MOMP) and attenuates apoptosis. Inhibits autophagy.
Specifications

Chemical Identifiers

338967-87-6
353.217
mdivi-1, mdivi 1, 3-2,4-dichloro-5-methoxyphenyl-2-sulfanyl-4 3h-quinazolinone, 3-2,4-dichloro-5-methoxyphenyl-2-sulfanylidene-1h-quinazolin-4-one, 3-2,4-dichloro-5-methoxyphenyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone, mdivi hplc , powder, 3-2,4-dichloro-5-methoxyphenyl-2-mercaptoquinazolin-4 3h-one, 3-2,4-dichloro-5-methoxyphenyl-2-sulfanylquinazolin-4-one, 3-2,4-dichloro-5-methoxyphenyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone, 3-2,4-dichloro-5-methoxyphenyl-2-thioxo-2,3-dihydro-4 1h-quinazolinone
3-(2,4-dichloro-5-methoxyphenyl)-2-sulfanylidene-1H-quinazolin-4-one
C15H10Cl2N2O2S
NZJKEVWTYMOYOR-UHFFFAOYSA-N
3825829
COC1=C(C=C(C(=C1)N2C(=O)C3=CC=CC=C3NC2=S)Cl)Cl

Specifications

Dynamin Inhibitors
338967-87-6
mdivi-1, mdivi 1, 3-2,4-dichloro-5-methoxyphenyl-2-sulfanyl-4 3h-quinazolinone, 3-2,4-dichloro-5-methoxyphenyl-2-sulfanylidene-1h-quinazolin-4-one, 3-2,4-dichloro-5-methoxyphenyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone, mdivi hplc , powder, 3-2,4-dichloro-5-methoxyphenyl-2-mercaptoquinazolin-4 3h-one, 3-2,4-dichloro-5-methoxyphenyl-2-sulfanylquinazolin-4-one, 3-2,4-dichloro-5-methoxyphenyl-2,3-dihydro-2-thioxo-4 1h-quinazolinone, 3-2,4-dichloro-5-methoxyphenyl-2-thioxo-2,3-dihydro-4 1h-quinazolinone
COC1=C(C=C(C(=C1)N2C(=O)C3=CC=CC=C3NC2=S)Cl)Cl
353.217
353.22
50mg
C15H10Cl2N2O2S
NZJKEVWTYMOYOR-UHFFFAOYSA-N
3-(2,4-dichloro-5-methoxyphenyl)-2-sulfanylidene-1H-quinazolin-4-one
3825829
>98%
Videos