Learn More
Tocris Bioscience™ (±)-McN 5652
Potent, orally active 5-HT uptake inhibitor. Also inhibits noradrenalin and dopamine uptake in vitro
Brand:  Tocris Bioscience™ 214850
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13430664
  • £367.04 / 50mg
Estimated Shipment
14-09-2020
Description
Description
Potent, high affinity serotonin re-uptake inhibitor; selective in vivo. Displays moderate selectivity over noradrenalin and dopamine re-uptake in vitro (Ki values are 0.68, 2.9 and 36.8 nM for inhibition of serotonin, noradrenalin and dopamine uptake respectively in rat brain synaptosomes). Potently potentiates 5-HT-induced effects in vivo following oral administration. Biological activity resides predominantly in the (+)-enantiomer.
Specifications
Specifications
(±)-McN 5652
>95
50mg
(±)-McN 5652
295.44