missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549747
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total MCM5 protein.
MCM5 Polyclonal antibody specifically detects MCM5 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
MCM5 | |
Polyclonal | |
Unconjugated | |
MCM5 | |
AA617332; AI324988; AL033333; Cdc46; CDC46 homolog; DNA replication licensing factor MCM5; MCM5; MCM5 minichromosome maintenance deficient 5, cell division cycle 46; Mcmd5; MGC5315; minichromosome maintenance complex component 5; minichromosome maintenance deficient 5 (cell division cycle 46); minichromosome maintenance deficient 5, cell division cycle 46; minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae); P1-CDC46 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17218, 291885, 4174 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P33992, P49718 | |
MCM5 | |
Synthesized peptide derived from human MCM5 antibody. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |