missing translation for 'onlineSavingsMsg'
Learn More

MCL-1, Alexa Fluor 488, clone: LVUBKM, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-9047-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510267

  • £87.34 / 25 tests


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This LVUBKM monoclonal antibody recognizes human and mouse myeloid cell leukemia sequence 1 (Mcl-1). Mcl-1 is an anti-apoptotic member of the Bcl-2 family of proteins important for regulation of cell survival/apoptosis. Mcl-1 is primarily localized to the outer membrane of mitochondria where it prevents cytochrome c release via dimerization with other Bcl-2 family members such as Bim. Although it is expressed in both immune and non-immune cells, highest levels of Mcl-1 expression are seen in hematopoietic lineage cells. PI3K activation of AKT results in destabilization and degradation of GSK3 beta, which prevents phosphorylation of Mcl-1 on S159 and its subsequent ubiquitnation and degradation. Mice conditionally lacking Mcl-1 in lymphocytes showed that Mcl-1 is essential during early lymphoid development and for the maintenance of mature lymphocytes.Applications Reported: This LVUBKM antibody has been reported for use in intracellular staining followed by flow cytometric analysis.Applications Tested: This LVUBKM antibody has been pre-titrated and tested by intracellular staining followed by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Staining Protocol: All protocols work well for this monoclonal antibody. Use of Protocol A: Two-step protocol: intracellular (cytoplasmic) proteins allows for the greatest flexibility for detection of surface and intracellular (cytoplasmic) proteins. Use of Protocol B: One-step protocol: intracellular (nuclear) proteins is recommended for staining of transcription factors in conjunction with surface and phosphorylated intracellular (cytoplasmic) proteins. Protocol C: Two-step protocol: Fixation/Methanol allows for the greatest discrimination of phospho-specific signaling between unstimulated and stimulated samples, but with limitations on the ability to stain specific surface proteins (refer to Clone Performance Following Fixation/Permeabilization located in the Best Protocols Section under the Resources tab online). All Protocols can be found in the Flow Cytometry Protocols: Staining Intracellular Antigens for Flow Cytometry Protocol located in the Best Protocols Section under the Resources tab online.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Mcl-1
LVUBKM
Alexa Fluor 488
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry (Intracellular Staining)
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
myeloid cell leukemia sequence 1
IgG1
RUO
Primary
Murine
Videos