missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBL-2 Rat anti-Mouse, Clone: 272801, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | MBL-2 |
---|---|
Clone | 272801 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse MBL-2 Glu19-Asp244 Accession # P41317 |
Gene ID (Entrez) | 17195 |
Gene Accession No. | P41317 |
Description
MBL-2 Monoclonal specifically detects MBL-2 in Mouse samples. It is validated for Western Blot.Specifications
MBL-2 | |
Mouse myeloma cell line NS0-derived recombinant mouse MBL-2 Glu19-Asp244 Accession # P41317 | |
P41317 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Rat | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
RUO |
272801 | |
17195 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse MBL-2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human MBL is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only