Learn More
Thermo Scientific™ Maxima™ H Minus cDNA Synthesis Master Mix, with dsDNase
Provide one-tube master mix for convenient and consistent cDNA synthesis for two-step RT-qPCR applications. The format helps streamline the RT-qPCR workflow, saves time, reduces pipeting errors to give customers sensitive, reliable and fast cDNA synthesis.
£373.00 - £1194.00
Specifications
For Use With (Application) Real-Time PCR
View More Specs
Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
15696019 View Documents Thermo Scientific™
M1681
50 £373.00
50 reactions View alternative products
Save Up to 
»
EU Stock 10 Please to purchase this item. Need a web account? Register with us today!
15606029 View Documents Thermo Scientific™
M1682
200 £1194.00
200 reactions View alternative products
Save Up to 
»
EU Stock 15 Please to purchase this item. Need a web account? Register with us today!
Description
Description
Thermo Scientific Maxima H Minus Reverse Transcriptase (RT), an engineered RNase H minus enzyme, is formulated into convenient one-tube master mix with a simplified protocol, enabling consistency and maximum control of RT-qPCR. It was developed through molecular evolution to improve thermostability, robustness and synthesis rates. Maxima H Minus RT also shows up to a 50x increase in processivity, has no RNase H activity, and demonstrates higher sensitivity.
  • Efficiently transcribes large and small amounts of RNA
  • Quick and convenient setup eliminates pipeting caused variability
  • Reduces risk of cross-contamination between samples
  • Consistently higher sensitivity for wide variety of gene targets
  • High linearity in cDNA synthesis over a broad dynamic range
  • dsDNase for fast and more efficient gDNA removal in RNA prep and no-RT control for RT-qPCR
  • dsDNase treated RNA samples show no decrease in RNA integrity or quantity
  • Complete enzyme activity from 42° to 55°C
Specifications
Specifications
Real-Time PCR