missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Matrilin-4 Antibody, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | Matrilin-4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Matrilin-4 Polyclonal specifically detects Matrilin-4 in Mouse samples. It is validated for Western Blot.Specifications
Matrilin-4 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
FLJ14417, MATN4, matrilin 4, matrilin-4, Matrilin4 | |
MATN4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O89029 | |
8785 | |
Mouse myeloma cell line NS0-derived recombinant mouse Matrilin-4 Gln22-Lys624 Accession # O89029 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Matrilin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Matrilin-4 is observed and less than 2% cross-reactivity with recombinant mouse Matrilin-2 and rhMatrilin-3 is observed. |
For Research Use Only