missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MARK2 Recombinant Human Protein
MARK2 is a serine/threonine protein kinase.
£1602.00 - £6900.00
Specifications
Accession Number | NP_059672.2 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 110.6kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438583
|
Invitrogen™
PR7405A |
100μg |
£1602.00
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10016942
|
Invitrogen™
PV4196 |
1mg |
£6900.00
1mg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MARK2 is involved in the control of cell polarity and microtubule stability.
Specifications
NP_059672.2 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
110.6kDa | |
RUO - research use only | |
MARK2, EMK1, PAR1 | |
Human | |
GST Tag | |
Frozen |