Learn More
Ansell Edmont™ Marigold™ Blue Natural Rubber Latex Immersion Gloves
Designed with high flexibility and excellent dexterity for chemical and liquid protection.
Brand:  Ansell Edmont™ A029070
Code : NEW
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 15356484
  • £62.67 / 12 pairs
In Stock 8
Add to basket
Description
Description
  • Crinkle finish provides grip in wet and dry environments
  • Polyamide seamless liner
  • Wrinkle natural rubber latex coating
  • Gauntlet cuff
Specifications
Specifications
Natural Rubber Latex
Natural Rubber Latex
No
Chemical and liquid
Astroflex
50mil
Immersion Gloves
Natural Rubber Latex
Knitted Elastic cuff
Natural Rubber Latex
7
144 Pairs
Standards EN 374, EN 407, EN 388 : 2241, 21 CFR (USA), CFIA (Canada)
Videos