missing translation for 'onlineSavingsMsg'
Learn More

Marginal Zone B Cells, clone: HIS57, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9489-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15567596

  • £154.20 / 100µg


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The monoclonal antibody HIS57 recognizes an unknown antigen expressed by rat marginal zone B cells. Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low.Applications Reported: This HIS57 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections.Applications Tested: This HIS57 antibody has been tested by immunohistochemistry of frozen rat tissue and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Marginal Zone B Cell Marker
HIS57
Unconjugated
Purified
Murine
100μg
Store at 2-8°C.
Monoclonal
Immunohistochemistry (Frozen)
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MZB1
IgG1
RUO
Primary
Rat
Videos