Learn More
Marginal Zone B Cells, clone: HIS57, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9489-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low.
Description: The monoclonal antibody HIS57 recognizes an unknown antigen expressed by rat marginal zone B cells. Marginal zone B cells are an anatomical B cell compartment located in the spleen at the area between red and white pulp. Marginal zone B cells serve as a first line of defense against blood-borne antigens in a T cell-independent manner. Marginal zone B cells can also be phenotyped as B cells that are IgM high, IgD low, CD21 high and CD23 low.Applications Reported: This HIS57 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of frozen tissue sections.Applications Tested: This HIS57 antibody has been tested by immunohistochemistry of frozen rat tissue and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.Specifications
Marginal Zone B Cells | |
Monoclonal | |
0.5mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
25 μg | |
Primary | |
Store at 2-8°C. | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
HIS57 | |
Unconjugated | |
MZB1, MZ-B | |
Affinity chromatography | |
RUO | |
Rat | |
Antibody | |
IgG1 κ |
For Research Use Only.