missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Marapsin/Pancreasin Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | Marapsin/Pancreasin |
---|---|
Clone | 243305 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Marapsin/Pancreasin Monoclonal specifically detects Marapsin/Pancreasin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Marapsin/Pancreasin | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
Q9BQR3 | |
83886 | |
Mouse myeloma cell line NS0-derived recombinant human Marapsin/Pancreasin Ala23-Lys290 Accession # Q9BQR3 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Marapsin/Pancreasin in direct ELISAs and Western blots. Reacts with an epitope within the mature chain. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Marapsin is observed. |
243305 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CAP-2, CAPH2, channel-activating protease 2, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, EC 3.4.21.43, marapsin, MPN, MPNMarapsin, Pancreasin, protease, serine 27, PRSS27, serine protease 27 | |
PRSS27 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only