missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP4K1 (HPK1), GST Tag Recombinant Protein
Human MAP4K1 (HPK1), GST Tag Recombinant Protein
£439.00 - £6540.00
Specifications
Molecular Weight (g/mol) | 65.0 kDa |
---|---|
Gene Alias | MAP4K1 |
Protein Length | aa 1-346 |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402172
|
Invitrogen™
PV6356 |
100 μg |
£1484.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15462870
|
Invitrogen™
PV6355 |
10μg |
£439.00
10µg |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15428993
|
Invitrogen™
PV6357 |
1mg |
£6540.00
1mg |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP4K1 is a tissue-specific upstream activator of the MEKK/JNK/SAPK signaling pathway. MAP4K1 diminishes T cell receptor (TCR) signaling activity and T cell proliferation by phosphorylating the adaptor protein SLP-76.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
65.0 kDa | |
aa 1-346 | |
Recombinant | |
Serine/Threonine Kinase | |
Kinases & Inhibitors | |
Liquid |
MAP4K1 | |
Human | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.25 mM DTT, 10 mM glutathione, 0.1 mM EDTA, 0.1 mM PMSF, 25% glycerol Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. |
|
Baculovirus | |
Recombinant, Full Length |