missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MAP3K14 (NIK), GST Tag Recombinant Protein
Human MAP3K14 (NIK), GST Tag Recombinant Protein
£430.00 - £6900.00
Specifications
Accession Number | NP_003945.2 |
---|---|
Gene Alias | MAP3K14 (NIK) |
KinaseFamily | STE Unique Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488693
|
Invitrogen™
PR8920A |
100μg |
£1588.00
100µg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478773
|
Invitrogen™
PV4903 |
1mg |
£6900.00
1mg |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307993
|
Invitrogen™
PV4902 |
10 μg |
£430.00
Each |
Estimated Shipment: 16-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes.
Specifications
NP_003945.2 | |
STE Unique Kinase Family | |
Recombinant | |
Baculovirus |
MAP3K14 (NIK) | |
Human | |
GST Tag |