missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2 Mouse, eFluor 570, Clone: AP20, eBioscience™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 41-9763-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
The monoclonal antibody AP20 recognizes the high molecular weight isoforms of microtubule-associated protein-2 (MAP2a and MAP2b) of human, rat, mouse, bovine, chicken, and xenopus. MAP2 belongs to a family of heat-stable phosphoproteins that are differentially expressed during brain development. MAP2 expression is localized to the cell body and dendrites of neurons and is involved in stabilizing microtubules by crosslinking with intermediate filaments. Expression of MAP2 is commonly used as a neural marker given that its expression becomes more pronounced with neuronal maturation.Specifications
MAP2 | |
AP20 | |
eFluor 570 | |
Conjugated | |
Murine | |
25μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Monoclonal |
Microscopy | |
0.2mg/mL | |
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer | |
Microtubule-associated protein 2 | |
IgG1, kappa | |
RUO | |
Primary | |
Xenopus |
For Research Use Only.