Learn More
MAP2 Mouse, Clone: AP20, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 14-9763-80
Code : NEW
Additional Details : Weight : 0.09500kg
 View more versions of this product
Product Code. 15507656
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Disclaimers:
For Research Use Only.
Description
Description
The monoclonal antibody AP20 recognizes the high molecular weight isoforms of microtubule-associated protein-2 (MAP2a and MAP2b) of human, rat, mouse, bovine, chicken, and xenopus. MAP2 belongs to a family of heat-stable phosphoproteins that are differentially expressed during brain development. MAP2 expression is localized to the cell body and dendrites of neurons and is involved in stabilizing microtubules by crosslinking with intermediate filaments. Expression of MAP2 is commonly used as a neural marker given that its expression becomes more pronounced with neuronal maturation.
Specifications
Specifications
MAP2
AP20
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Microscopy
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Microtubule-associated protein 2
IgG1
RUO
Primary
Xenopus
Videos