missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mannitol, Extra Pure, SLR, Fisher Chemical
Click to view available options
500g, Plastic powder jar
Specifications
CAS | 69-65-8 |
Melting Point | 165°C |
Color | White |
Packaging | Plastic powder jar |
Molecular Formula | C6H14O6 |
MDL Number | 64287 |
Quantity | 500 g |
Formula Weight | 182.17g/mol |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'