missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganous Chloride, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
267.19 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Specifications
Physical Form | Crystals |
Quantity | 500 g |
Formula Weight | 197.9 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.01kg |
Chemical Name or Material | Manganous Chloride, 4-Hydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'