missing translation for 'onlineSavingsMsg'
Learn More

Manganese sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis), Thermo Scientific™

Catalog Number 15494795
Click to view available options
Quantity:
1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

Magnetron sputtering source

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 7439-96-5
Molecular Formula Mn
MDL Number MFCD00011111
Quantity 1 Ea.
Synonym colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
Solubility Information Soluble in diluted acids. Insoluble in water.
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
SMILES [Mn]
IUPAC Name manganese
Molecular Weight (g/mol) 54.94
PubChem CID 23930
ChEBI CHEBI:35154
Percent Purity 99.95%
Odor Odorless
Form Hot Pressed
Assay (metals basis)
Chemical Name or Material Manganese sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'