missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(Iv) Oxide, Extra Pure, SLR, Precipitated, Fisher Chemical™
Click to view available options
Quantity:
500 g
Specifications
Melting Point | 535°C |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 86.94g/mol |
Packaging | Glass Jar |
Safety and Handling
Product Identifier
- Manganese(IV) oxide, electrolytically precipitated, active
- Danger
- Acute toxicity Category 4
- Oxidising solid Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'