missing translation for 'onlineSavingsMsg'
Learn More

Manganese(III) acetate meso-tetraphenylporphine, Thermo Scientific™

Catalog Number p-7050707
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
58356-65-3
C46H32MnN4O2
727.726
MFCD00216548
UFMFSWMECMUSKE-UHFFFAOYSA-N
manganese iii acetate meso-tetraphenylporphine, manganese meso-tetraphenylporphine acetate, manganese iii meso-tetraphenylporphine acetate, 2,7,12,17-tetraphenyl-21,23,24,25-tetraaza-22-manganahexacyclo 9.9.3.1^ 3,6 .1^ 13,16 .0^ 8,23 .0^ 18,21 pentacosa-1,3 25 ,4,6,8,10,12,14,16 24 ,17,19-undecaene; acetic acid
44135862
acetic acid;manganese(2+);5,10,15,20-tetraphenylporphyrin-22,24-diide
CC(=O)O.C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C=C4)C9=CC=CC=C9)[N-]3.[Mn+2]
This item is not returnable. View return policy
58356-65-3
C46H32MnN4O2
727.726
MFCD00216548
UFMFSWMECMUSKE-UHFFFAOYSA-N
manganese iii acetate meso-tetraphenylporphine, manganese meso-tetraphenylporphine acetate, manganese iii meso-tetraphenylporphine acetate, 2,7,12,17-tetraphenyl-21,23,24,25-tetraaza-22-manganahexacyclo 9.9.3.1^ 3,6 .1^ 13,16 .0^ 8,23 .0^ 18,21 pentacosa-1,3 25 ,4,6,8,10,12,14,16 24 ,17,19-undecaene; acetic acid
44135862
acetic acid;manganese(2+);5,10,15,20-tetraphenylporphyrin-22,24-diide
CC(=O)O.C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C=C4)C9=CC=CC=C9)[N-]3.[Mn+2]

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'