missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) sulfate monohydrate, Puriss., meets analytical spec. of Ph. Eur, BP, USP, FCC, 99 to 100.5% (calc. for dried substance), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
2.5 kg
50 kg
Unit Size:
1kg
2.5kg
50kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 1 kg |
| Linear Formula | MnSO4 · H2O |
| UN Number | 3077 |
| Formula Weight | 169.02g/mol |
| Percent Purity | 99 to 100.5% (calc. to the dried substance) |
| Grade | puriss. |
| Chemical Name or Material | Manganese(Ii) Sulfate Monohydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction