missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog Number 10507562
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
10034-96-5
H2MnO5S
169.01
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
10034-96-5
H2MnO5S
169.01
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 850.0°C
Melting Point 700.0°C
Color Pink
Physical Form Powder
Quantity 5 g
Assay Percent Range 98.0 to 101% (Complexometry)
Linear Formula MnSO4·H2O
Merck Index 15, 5804
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
IUPAC Name manganese(2+);sulfate;hydrate
Formula Weight 169.02
Percent Purity ≥98%
Grade ACS Reagent
Loss on Ignition 10.0% to 12.0%
Packaging Glass Bottle
Chemical Name or Material Manganese(II) sulfate monohydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) sulfate monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'