missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) oxalate dihydrate, Mn 30% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
50 g
250 g
1 kg
Description
Manganese(II) oxalate dihydrate is used as a paint and varnish drier. It is also used as a chemical reagent as well as to prepare other manganese compounds. In addition, it is used as a semiconductor photosensitive material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 100°C (decomposition) |
Color | White |
Density | 2.453 g/mL |
Odor | Odorless |
Quantity | 1 kg |
Merck Index | 14,5734 |
Solubility Information | Soluble in dilute acids; Slightly soluble in water. |
Formula Weight | 178.99 (142.96 Anhydrous) |
Concentration or Composition (by Analyte or Components) | Mn 30% min. |
Physical Form | Chunks and/or Powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'