missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Manganese(II) naphthenate, 56% w/w in mineral spirits, 6-10% Mn

Brand:  Alfa Aesar™ 038609.22

Code : NEW

Additional Details : CAS Number : 1336-93-2 Weight : 0.10000kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 100g

 View more versions of this product

Product Code. 11398927

  • £83.63 / 100g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Specifications

Chemical Identifiers

Manganese(II) naphthenate
1336-93-2
MFCD00147533
UN1993

Specifications

Manganese(II) naphthenate
40° (104°F)
100g
Liquid
UN1993
56% w/w in min.eral spirits, 6 to 10% Mn
Mineral-oil-like
1336-93-2
MFCD00147533
Videos
SDS