missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11322418
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7773-01-5
Cl2Mn
125.84
MFCD00011114
GLFNIEUTAYBVOC-UHFFFAOYSA-L
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
10313134
CHEBI:63041
[Cl-].[Cl-].[Mn++]
This item is not returnable. View return policy
7773-01-5
Cl2Mn
125.84
MFCD00011114
GLFNIEUTAYBVOC-UHFFFAOYSA-L
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
10313134
CHEBI:63041
[Cl-].[Cl-].[Mn++]

Manganese(II) chloride is used in formation of an active Mn(0) species useful for radical cyclization reactions. It is used as catalyst, raw material for paint drier and antiknock agent for gasoline. It iis also used in the production of pigments, dyes, car batteries, chemical reagents and trace elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 1190°C
Melting Point 650°C
Color Pink
Physical Form Beads
Quantity 5 g
Linear Formula MnCl2
UN Number UN3288
Merck Index 14,5728
Solubility Information Soluble in water(723g/L at 25 C)
IUPAC Name manganese(2+);dichloride
Formula Weight 125.84
Percent Purity 99.99%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 2.977 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(II) chloride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'