missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(Ii) Chloride Tetrahydrate, 99.99% trace metals basis, Honeywell™
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Poly Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Melting Point | 58°C |
Color | Pink |
Quantity | 25 g |
Linear Formula | MnCl2 · 4H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 197.91g/mol |
Percent Purity | 99.99% (trace metals basis) |
Packaging | Poly Bottle |
Appearance | Crystals |
Density | 1.913g/cu.cm |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'