missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) carbonate, 99.9% (metals basis excluding Na), Thermo Scientific Chemicals

Catalog Number 11317907
Click to view available options
Quantity:
250 g
1 kg
598-62-9
CMnO3
114.946
MFCD00011116
XMWCXZJXESXBBY-UHFFFAOYSA-L
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
11726
manganese(2+);carbonate
C(=O)([O-])[O-].[Mn+2]
598-62-9
CMnO3
114.946
MFCD00011116
XMWCXZJXESXBBY-UHFFFAOYSA-L
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
11726
manganese(2+);carbonate
C(=O)([O-])[O-].[Mn+2]

Manganese(II) carbonate is used as an additive to plant fertilizers, glaze colorant in ceramics, pigment and drier for varnishes. It is also used as an ingredient in welding rod, animal feed additive as well as to prepare other manganese salts and pharmaceuticals. It is also used in medicine as a hematinic and in health foods, in flux and in concrete stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 350°C (decomposition)
Color Brown to Violet
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.9% (metals basis excluding Na)
Solubility Information Soluble in dilute inorganic acids and carbon dioxide. Insoluble in water,alcohol,ammonia and common organic acids.
Formula Weight 114.95
Odor Odorless
Appearance Light brown to violet powder
Density 3.125 g/cm3
Chemical Name or Material Manganese(II) carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'