missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide, 99%, anhydrous, Thermo Scientific Chemicals

Catalog Number p-3776960
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
13446-03-2
Br2Mn
214.76
MFCD00011113
13446-03-2
Br2Mn
214.76
MFCD00011113

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Pink
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Linear Formula MnBr2
Merck Index 15, 5790
Solubility Information Solubility in water: soluble
Formula Weight 214.76
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 4.38
Density 4.3800g/mL
Chemical Name or Material Manganese(II) bromide, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Manganese(II) bromide, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'