missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate, 98+%, anhydrous, Thermo Scientific Chemicals

Catalog Number 10540374
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
638-38-0
C4H6MnO4
173.03
MFCD00013039
UOGMEBQRZBEZQT-UHFFFAOYSA-L
manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y
12525
manganese(2+);diacetate
[Mn++].CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
638-38-0
C4H6MnO4
173.03
MFCD00013039
UOGMEBQRZBEZQT-UHFFFAOYSA-L
manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y
12525
manganese(2+);diacetate
[Mn++].CC([O-])=O.CC([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Beige-Gray to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula Mn(OC(O)CH3)2
Merck Index 15, 5789
Solubility Information Solubility in water: very soluble
Formula Weight 173.03
Percent Purity ≥98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Manganese(II) acetate
Show More Show Less
Product Identifier
  • Manganese(II) acetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'