missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn, Thermo Scientific Chemicals
14.73 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
Used as paint driers. Accelerating curing agent for unsaturated polyester. Also used as a intermediate for dyestuff, pharmaceutical, syntheses material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
Density | 0.897 g/mL |
Flash Point | 40°C (104°F) |
Odor | Petroleum-like |
Quantity | 250 g |
UN Number | UN1268 |
Solubility Information | Soluble in organic solvents. Not miscible or difficult to mix with water. |
Formula Weight | 341.35 |
Concentration or Composition (by Analyte or Components) | 40% w/w in min.eral spirits, 6% Mn |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'