missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Malt Extract NPS Including Dispenserfilter

Sterile, dehydrated culture media pads with a gridded membrane filter.

Brand:  Sartorius Stedim Biotech™ 14086--50----RDN

Additional Details : Weight : 1.16500kg

 View more versions of this product

Product Code. 15952500

  • £202.98 / Pack of 100

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

  • Malt extract nutrient pads with gridded membranes
  • For determination of yeasts and molds acc. to international Standards e.g. APHA, AOAC, IFU and internal SOPs.
  • Trouble-free storage up to 24 months
  • Economical by time- and cost-savings
  • Highly versatile -multiple applications
  • Consistent quality
  • Validation guide available
  • Easy handling
  • Compliance with international standards and references
Specifications

Specifications

10 x 10 petri dishes with NPS in bags, dispenser-ready membrane filters
Malt Extract
Gray
White
Yeasts and Molds
0.8μm Pore Size Disc
Sterile
50 mm
0.8μm
100/Pk.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers