missing translation for 'onlineSavingsMsg'
Learn More

Malonyl dichloride, 97%, ACROS Organics™

Brand:  Acros Organics 125281000

Code : 29

Additional Details : CAS Number : 1663-67-8 Weight : 0.26000kg Transport : UN number : 2920 Chem class : 1a Pack group : II

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10692135

  • £203.70 / 100g


In Stock 1
Add to basket

Specifications

Chemical Identifiers

Malonyl dichloride
97%
ClCOCH2COCl
MFCD00000735
97%
C3H2Cl2O2
140.95

Specifications

Malonyl dichloride
1.45
Clear yellow to orange to brown
1663-67-8
Liquid
97%
Authentic
140.95
02, I, 252
1.4620 to 1.4660
47°C
47.0°C (15.0 mmHg)
100g
1.4500g/mL
97%
C3H2Cl2O2
ClCOCH2COCl
MFCD00000735
12,291
Glass bottle
Videos
SDS