missing translation for 'onlineSavingsMsg'
Learn More

Malonic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-7081518
21.76 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2500 g
141-82-2
C3H4O4
104.061
MFCD00002707
OFOBLEOULBTSOW-UHFFFAOYSA-N
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
867
CHEBI:30794
propanedioic acid
C(C(=O)O)C(=O)O
141-82-2
C3H4O4
104.061
MFCD00002707
OFOBLEOULBTSOW-UHFFFAOYSA-N
malonic acid, dicarboxymethane, carboxyacetic acid, methanedicarboxylic acid, kyselina malonova, usaf ek-695, dicarboxylate, dicarboxylic acid, kyselina malonova czech, propanediolic acid
867
CHEBI:30794
propanedioic acid
C(C(=O)O)C(=O)O

Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonic acid is acts as a building block in organic synthesis. It is also useful as a precursor for polyesters and alkyd resins, which is used in coating applications, thereby protecting against UV light, corrosion and oxidation. It acts as a cross linker in the coating industry and surgical adhesive. It finds application in the production of specialty chemicals, flavors and fragrances, polymer cross linkers and pharmaceuticals.

Solubility
Soluble in cold water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 132°C to 136°C (decomposition)
Density 1.62
Flash Point 157°C (314°F)
Quantity 100 g
Beilstein 1751370
Merck Index 14,5710
Solubility Information Soluble in cold water.
Formula Weight 104.06
Percent Purity 99%
Chemical Name or Material Malonic acid
compliance-icons
Product Identifier
  • Malonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'