missing translation for 'onlineSavingsMsg'
Learn More

Malonaldehyde bis(dimethyl acetal), 99+%, Thermo Scientific Chemicals

Catalog Number 13498368
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
102-52-3
C7H16O4
164.20
MFCD00008488
XHTYQFMRBQUCPX-UHFFFAOYSA-N
propane, 1,1,3,3-tetramethoxy, tetramethoxypropane, malonaldehyde bis dimethyl acetal, malonaldehyde tetramethyl acetal, malonaldehyde, bis dimethyl acetal, 1,1,3,3-tetramethoxy-propane, malonaldehyde bis dime, 1,3,3-tetramethoxypropane, ksc174o0b, propane,1,3,3-tetramethoxy
66019
1,1,3,3-tetramethoxypropane
COC(CC(OC)OC)OC
This item is not returnable. View return policy
102-52-3
C7H16O4
164.20
MFCD00008488
XHTYQFMRBQUCPX-UHFFFAOYSA-N
propane, 1,1,3,3-tetramethoxy, tetramethoxypropane, malonaldehyde bis dimethyl acetal, malonaldehyde tetramethyl acetal, malonaldehyde, bis dimethyl acetal, 1,1,3,3-tetramethoxy-propane, malonaldehyde bis dime, 1,3,3-tetramethoxypropane, ksc174o0b, propane,1,3,3-tetramethoxy
66019
1,1,3,3-tetramethoxypropane
COC(CC(OC)OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.9900g/mL
Boiling Point 183.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Linear Formula (CH3O)2CHCH2CH(OCH3)2
Refractive Index 1.4060 to 1.4080
Quantity 5 kg
Specific Gravity 0.99
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in hexane,ether,alcohol
Formula Weight 164.2
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Malonaldehyde bis(dimethyl acetal), 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Malonaldehyde bis(dimethyl acetal)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'