missing translation for 'onlineSavingsMsg'
Learn More

Maleimide, 98%, Thermo Scientific Chemicals

Catalog Number 10633192
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
541-59-3
C8H4AgKN2O4
339.10
MFCD00005494
KWLWLQRUFFGWNG-UHFFFAOYSA-L
maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one
10935
CHEBI:16072
[K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1
This item is not returnable. View return policy
541-59-3
C8H4AgKN2O4
339.10
MFCD00005494
KWLWLQRUFFGWNG-UHFFFAOYSA-L
maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one
10935
CHEBI:16072
[K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 90.0°C to 94.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21,399
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethylether
IUPAC Name pyrrole-2,5-dione
Formula Weight 97.07
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Maleimide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Maleimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'