missing translation for 'onlineSavingsMsg'
Learn More

Maleic Hydrazide, Single-Component Organic Standards, SPEX CertiPrep™

Brand:  SPEX Certiprep S2357

UNSPSC : 12350000

Code : 04

Additional Details : Weight : 0.10000kg

Quantity 1mL
Packaging 2mL amber prescored glass ampule

 View more versions of this product

Product Code. 12909284

  • £49.00 / 1mL


Call for latest availability
+44 (0)1509 555 500
Add to basket

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

AnalyteCAS No.Concentration
Maleic anhydride108-31-61000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

Maleic anhydride
67-56-1
99.9
99.9

Specifications

Maleic anhydride
99.9
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
1mL
67-56-1
99.9
For US EPA Method 8270
Methanol-P&T
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation