missing translation for 'onlineSavingsMsg'
Learn More

Maleate, 0.2M buffer soln., pH 6.0, Thermo Scientific™

Catalog Number 15494489
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
3105-55-3
C4H2NaO4
137.05
MFCD00070246
VRVKOZSIJXBAJG-ODZAUARKSA-L
91658932
[Na+].[O-]C(=O)\C=C/C([O-])=O
This item is not returnable. View return policy
3105-55-3
C4H2NaO4
137.05
MFCD00070246
VRVKOZSIJXBAJG-ODZAUARKSA-L
91658932
[Na+].[O-]C(=O)\C=C/C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.2 M
pH 6.0
Physical Form Liquid
Quantity 250 mL
IUPAC Name sodium (2Z)-but-2-enedioate
Chemical Name or Material Maleate buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'