missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAGP-2/MFAP5 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | MAGP-2/MFAP5 |
---|---|
Applications | Western Blot |
Conjugate | Unconjugated |
Host Species | Sheep |
Regulatory Status | RUO |
Description
MAGP-2/MFAP5 Polyclonal specifically detects MAGP-2/MFAP5 in Human samples. It is validated for Western Blot.Specifications
MAGP-2/MFAP5 | |
Unconjugated | |
RUO | |
8076 | |
Polyclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
Sheep | |
Human | |
Mouse myeloma cell line NS0-derived recombinant human MAGP-2/MFAP5 Ile22-Leu173 Accession # Q13361 | |
Primary |
For Research Use Only