Learn More
Invitrogen™ MagniSort™ Streptavidin Positive Selection Beads
Designed for the magnetic separation of cells by positive selection. Desired cells are bound by biotinylated antibody and then streptavidin-coated magnetic beads. When placed in a magnetic field, desired cells are held in place and undesired cells can be separated by decanting.
Brand:  Invitrogen™ MSPB-6003-74
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15592567
  • £203.00 / 200 tests
Estimated Shipment
13-05-2021
Add to basket
Alert:
Contains Sodium azide
Description
Description
  • Clone and concentration of biotinylated antibody for desired cell labeling, as well as concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically
  • Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads
  • The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody
  • For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol
Specifications
Specifications
200 tests
Streptavidin
Biotin