missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MagniSort™ Streptavidin Negative Selection Beads
Designed for the magnetic separation of cells by negative selection. Undesired cells are bound by a cocktail of biotinylated antibodies and then streptavidin coated magnetic beads.
£68.25 - £258.00
Specifications
Form | Liquid |
---|---|
Ligand | Streptavidin |
Product Type | Streptavidin Conjugate (Fluorescent) |
Target Molecule | Biotin |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15572567
|
Invitrogen™
MSNB-6002-74 |
200 Tests |
£258.00
200 tests |
In Stock
Log in to see stock. |
|||||
15562567
|
Invitrogen™
MSNB-6002-71 |
25 Tests |
£68.25
25 tests |
Estimated Shipment: 10-04-2024
Log in to see stock. |
|||||
Description
- Clone and concentration of biotinylated antibody for undesired cell labeling, as well as concentration of MagniSort™ Streptavidin Negative Selection Beads must be determined empirically
- Please refer to the protocol for details on how to optimize both the antibody or antibody cocktail and MagniSort™ Streptavidin Negative Selection Beads.
- For depletion of cells using a single antibody, we recoomend the MagniSort™ Streptavidin Positive Selection Beads (cat. MSPB-6003) and protocol
Specifications
Liquid | |
Streptavidin | |
Streptavidin Conjugate (Fluorescent) | |
Biotin |
For Research Use Only. Not for use in diagnostic procedures.