Learn More
Kartell™ Magnetic Triangular Stirring Bars
Made from Polytetrafluoroethylene. Kartell™ 120° Magnetic Triangular Stirring Bars are ideal for dissolving solids and mixing sediments thanks to the scraping action. The angled surfaces create high turbulence even at low speeds.
Brand: Kartell™ 0078100
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Cuneiform
Angled surface
Suitable for foodstuff
Magnet PTFE coated
Specifications
7.39x25.x32mm | |
PTFE (Polytetrafluoroethylene) |